This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198805_g1
See other DGKQ GE Assays ›
Gene Symbol:
Dgkq
Gene Name:
diacylglycerol kinase, theta
Gene Aliases:
110kDa, DAGK, DAGK7, Dagk4, Dgkd
Chromosome Location:
Chr.5: 108647044 - 108660828 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198805_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dgkq
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_199011.1 NP_950176.1 3 - 4 509 69
GenBank mRNA AK048764.1 - 3 - 4 551 69
AK170840.1 - 3 - 4 530 69
BC062929.1 - 3 - 4 509 6855058 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002952.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dgkq

Panther Classification:

Molecular Function -

kinase

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
signal transduction
G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
cAMP-mediated signaling
response to ATP
intracellular signal transduction
thrombin receptor signaling pathway
protein kinase C signaling
nucleotide binding
diacylglycerol kinase activity
ATP binding
kinase activity
transferase activity
kinase binding
activating transcription factor binding
phospholipase binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026147.mm.1 NM_199011.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026147.mm.1 NM_199011.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026147.mm.1 TC0500002952.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500026148.mm.1 NM_199011.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026148.mm.1 NM_199011.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026148.mm.1 TC0500002952.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0