This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01198463_m1
See other ALDH2 GE Assays ›
Gene Symbol:
Aldh2
Gene Name:
aldehyde dehydrogenase 2, mitochondrial
Gene Aliases:
AHD-M1, ALDH-E2, ALDHI, Ahd-5, Ahd5
Chromosome Location:
Chr.5: 121567687 - 121593824 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01198463_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Aldh2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK035145.1 - 4 - 5 549 90
AK043506.1 - 4 - 5 543 90
AK078626.1 - 4 - 5 549 90
AK079602.1 - 4 - 5 524 90
AK128913.1 - 4 - 5 486 90
AK146286.1 - 4 - 5 542 90
AK150233.1 - 4 - 5 541 90
AK150250.1 - 4 - 5 554 90
AK150920.1 - 4 - 5 541 90
AK150986.1 - 4 - 5 523 90
AK151763.1 - 4 - 5 541 90
AK151826.1 - 4 - 5 523 90
AK153128.1 - 4 - 5 523 90
AK160060.1 - 4 - 5 543 90
AK163452.1 - 549 90
AK165032.1 - 4 - 5 787 90
BC005476.1 - 4 - 5 531 3600875 90
S71509.1 - 4 - 5 505 90
U07235.1 - 4 - 5 548 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003193.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

dehydrogenase oxidoreductase metabolite interconversion enzyme

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500028451.mm.1 NM_009656.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500028451.mm.1 TC0500003193.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0500028450.mm.1 NM_009656.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500028450.mm.1 TC0500003193.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0