This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196636_g1
See other RGS14 GE Assays ›
Gene Symbol:
Rgs14
Gene Name:
regulator of G-protein signaling 14
Gene Aliases:
RPIP1
Chromosome Location:
Chr.13: 55369712 - 55384691 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196636_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rgs14
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016758.3 NP_058038.2 3 - 4 402 72
GenBank mRNA AK154944.1 - 3 - 4 327 72
BC030321.1 - 3 - 4 374 5374531 72
U85055.1 - 3 - 4 242 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000657.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rgs14

Gene Ontology Categories:

Function(s) Process(es)

nucleocytoplasmic transport
response to oxidative stress
cell cycle
spindle organization
chromosome segregation
mitotic nuclear division
signal transduction
G-protein coupled receptor signaling pathway
multicellular organism development
learning
long-term memory
regulation of G-protein coupled receptor protein signaling pathway
visual learning
negative regulation of signal transduction
zygote asymmetric cell division
negative regulation of synaptic plasticity
intracellular signal transduction
termination of G-protein coupled receptor signaling pathway
negative regulation of MAP kinase activity
positive regulation of GTPase activity
regulation of DNA-templated transcription in response to stress
platelet-derived growth factor receptor signaling pathway
positive regulation of neurogenesis
cell division
long-term synaptic potentiation
negative regulation of ERK1 and ERK2 cascade
G-protein alpha-subunit binding
receptor signaling protein activity
GDP-dissociation inhibitor activity
GTPase activator activity
protein binding
microtubule binding
protein kinase binding
receptor signaling complex scaffold activity
GTPase regulator activity
GTPase activating protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300004325.mm.1 NM_016758.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004325.mm.1 NM_016758.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004325.mm.1 TC1300000657.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004329.mm.1 NM_016758.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004329.mm.1 NM_016758.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004329.mm.1 TC1300000657.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004326.mm.1 NM_016758.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004326.mm.1 NM_016758.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004326.mm.1 TC1300000657.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004330.mm.1 NM_016758.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004330.mm.1 NM_016758.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004330.mm.1 TC1300000657.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300004328.mm.1 NM_016758.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300004328.mm.1 NM_016758.3 Refseq
Affymetrix Mouse Clariom S Array PSR1300004328.mm.1 TC1300000657.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0