This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196175_m1
See other ADRBK2 GE Assays ›
Gene Symbol:
Adrbk2
Gene Name:
adrenergic receptor kinase, beta 2
Gene Aliases:
4833444A01Rik, AI851927, AW551196, Adrbk-2, Bark-2, GRK3
Chromosome Location:
Chr.5: 112910478 - 113015951 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196175_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Adrbk2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285806.1 NP_001272735.1 8 - 9 748 80
NM_177078.4 NP_796052.2 8 - 9 709 80
GenBank mRNA AK076533.1 - 8 - 9 607 80
AK134672.1 - 8 - 9 686 80
AK141186.1 - 8 - 9 727 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003024.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Adrbk2

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of protein phosphorylation
desensitization of G-protein coupled receptor protein signaling pathway
protein phosphorylation
intracellular protein transport
signal transduction
phosphorylation
receptor internalization
termination of G-protein coupled receptor signaling pathway
inositol phosphate metabolic process
rhodopsin metabolic process
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
G-protein coupled receptor kinase activity
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
D1 dopamine receptor binding
beta-adrenergic receptor kinase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026881.mm.1 NM_001285806.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026881.mm.1 NM_177078.4 Refseq
Affymetrix Mouse Clariom S Array PSR0500026881.mm.1 TC0500003024.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0