This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01196014_g1
See other ANTXR2 GE Assays ›
Gene Symbol:
Antxr2
Gene Name:
anthrax toxin receptor 2
Gene Aliases:
2310046B19Rik, AW561899, CMG-2, CMG2, cI-35
Chromosome Location:
Chr.5: 97884688 - 98030962 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01196014_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Antxr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133738.1 NP_598499.1 5 - 6 900 80
GenBank mRNA AK155460.1 - 5 - 6 983 80
AK166124.1 - 672 80
AK170655.1 - 5 - 6 538 80
AK171292.1 - 5 - 6 575 80
BC076595.1 - 5 - 6 900 30649897 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500002805.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Antxr2

Panther Classification:

Molecular Function -

cell adhesion molecule

Gene Ontology Categories:

Function(s) Process(es)

reproductive process
toxin transport
receptor activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500024661.mm.1 NM_133738.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500024661.mm.1 NM_133738.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500024661.mm.1 TC0500002805.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-88d4p:80/100.66.130.94:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0