This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01195470_m1
See other GTF2IRD1 GE Assays ›
Gene Symbol:
Gtf2ird1
Gene Name:
general transcription factor II I repeat domain-containing 1
Gene Aliases:
1700012P16Rik, BEN, Cream1, ESTM9, GTF3, Gtf2il, MusTRD1, Tg(Alb1-Myc)166.8Sst, WBSCR11, X83320
Chromosome Location:
Chr.5: 134357661 - 134456716 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01195470_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gtf2ird1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081462.2 NP_001074931.1 16 - 17 1944 58
NM_001081463.2 NP_001074932.1 16 - 17 1944 58
NM_001081464.2 NP_001074933.1 16 - 17 1944 58
NM_001081465.1 NP_001074934.1 16 - 17 1887 58
NM_001081466.2 NP_001074935.1 16 - 17 1944 58
NM_001081467.2 NP_001074936.1 16 - 17 1944 58
NM_001081468.2 NP_001074937.1 16 - 17 1944 58
NM_001081469.2 NP_001074938.1 16 - 17 1944 58
NM_001081470.2 NP_001074939.1 16 - 17 1944 58
NM_001244936.1 NP_001231865.1 16 - 17 1944 58
NM_020331.3 NP_065064.2 16 - 17 1944 58
GenBank mRNA AF257475.1 - 16 - 17 1920 58
AF260133.1 - 16 - 17 1915 58
AF343348.1 - 16 - 17 1812 58
AF343349.1 - 16 - 17 1818 58
AF343350.1 - 16 - 17 1814 58
AF343351.1 - 16 - 17 1812 58
AF497637.1 - 16 - 17 1804 58
AF497638.1 - 16 - 17 1804 58
AF497639.1 - 16 - 17 1804 58
AF497640.1 - 16 - 17 1812 58
AF497641.1 - 16 - 17 1818 58
AF497642.1 - 16 - 17 1814 58
AK078488.1 - 16 - 17 1812 58
AK079487.1 - 16 - 17 1884 58
AK147196.1 - 16 - 17 1927 58
AY030287.1 - 16 - 17 1915 58
AY030288.1 - 16 - 17 1915 58
AY030289.1 - 16 - 17 1915 58
BC050822.1 - 16 - 17 1939 6393328 58
BC057091.1 - 16 - 17 1766 5703300 58
BC058964.1 - 16 - 17 1766 5703300 58
BC083150.1 - 16 - 17 1846 30459984 58

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gtf2ird1

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
multicellular organism development
embryo development
transition between slow and fast fiber
DNA binding
transcription factor activity, sequence-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0