This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194663_g1
See other PRIMPOL GE Assays ›
Gene Symbol:
Primpol
Gene Name:
primase and polymerase (DNA-directed)
Gene Aliases:
BC065112, Ccdc111
Chromosome Location:
Chr.8: 46575579 - 46617234 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194663_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Primpol
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001001184.1 NP_001001184.1 8 - 9 1103 142
GenBank mRNA BC065112.1 - 8 - 9 1103 30532532 142

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Primpol

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA replication
mitochondrial DNA replication
DNA replication, synthesis of RNA primer
DNA repair
transcription, DNA-templated
cellular response to DNA damage stimulus
response to UV
translesion synthesis
replication fork processing
chromatin binding
telomerase activity
RNA helicase activity
DNA-directed DNA polymerase activity
DNA primase activity
DNA-directed RNA polymerase activity
sulfate adenylyltransferase activity
RNA guanylyltransferase activity
tRNA guanylyltransferase activity
mannose-phosphate guanylyltransferase activity
transferase activity
nucleotidyltransferase activity
DNA/RNA helicase activity
CTP:2,3-di-O-geranylgeranyl-sn-glycero-1-phosphate cytidyltransferase activity
phospholactate guanylyltransferase activity
ATP:coenzyme F420 adenylyltransferase activity
UDP-N-acetylgalactosamine diphosphorylase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800016650.mm.1 NM_001001184.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800016650.mm.1 NM_001001184.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800016650.mm.1 TC0800002160.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0