This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194550_m1
See other SETD1B GE Assays ›
Gene Symbol:
Setd1b
Gene Name:
SET domain containing 1B
Gene Aliases:
AA516740, BC035291, KMT2G, mKIAA1076
Chromosome Location:
Chr.5: 123140787 - 123168634 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194550_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Setd1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040398.2 NP_001035488.2 11 - 12 4239 72
GenBank mRNA AK122435.1 - 3 - 4 574 72
BC035291.1 - 3 - 4 741 72
BC038367.1 - 5 - 6 1317 72
BC040775.1 - 3 - 4 741 72
BC041681.1 - 3 - 4 760 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500001410.mm.1

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

histone modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
methylation
nucleotide binding
molecular_function
nucleic acid binding
RNA binding
methyltransferase activity
transferase activity
histone-lysine N-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500012507.mm.1 NM_001040398.2 Refseq
Affymetrix Mouse Clariom S Array PSR0500012507.mm.1 TC0500001410.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0