This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01194122_m1
See other SGSM1 GE Assays ›
Gene Symbol:
Sgsm1
Gene Name:
small G protein signaling modulator 1
Gene Aliases:
2410098H20Rik, AA408781, D5Bwg1524e, E130107M15, NuIP, Rutbc2
Chromosome Location:
Chr.5: 113243220 - 113310787 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01194122_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sgsm1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162965.1 NP_001156437.1 9 - 10 1022 74
NM_001254731.1 NP_001241660.1 9 - 10 1022 74
NM_172718.3 NP_766306.2 9 - 10 1022 74
GenBank mRNA AK044716.1 - 9 - 10 976 74
AK053555.1 - 9 - 10 956 74
AK170021.1 - 9 - 10 1014 74
BC138049.1 - 9 - 10 948 8861068 74
BC138050.1 - 9 - 10 948 8861069 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0500003028.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Sgsm1

Panther Classification:

Molecular Function -

GTPase-activating protein

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_001162965.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_001162965.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_001254731.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_001254731.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_172718.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 NM_172718.3 Refseq
Affymetrix Mouse Clariom S Array PSR0500026976.mm.1 TC0500003028.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0