This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193947_g1
See other CABP7 GE Assays ›
Gene Symbol:
Cabp7
Gene Name:
calcium binding protein 7
Gene Aliases:
-
Chromosome Location:
Chr.11: 4738821 - 4746778 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193947_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cabp7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_138948.3 NP_620398.1 3 - 4 419 65
GenBank mRNA AF419324.1 - 3 - 4 420 65
BC104356.1 - 3 - 4 418 40049623 65
BC104357.1 - 3 - 4 418 40049625 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002120.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cabp7

Gene Ontology Categories:

Function(s) Process(es)

biological_process
calcium ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100020584.mm.1 NM_138948.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100020584.mm.1 NM_138948.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100020584.mm.1 TC1100002120.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0