This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01193675_m1
See other TRAF3IP2 GE Assays ›
Gene Symbol:
Traf3ip2
Gene Name:
TRAF3 interacting protein 2
Gene Aliases:
AI429613, Act1, CIKS
Chromosome Location:
Chr.10: 39612934 - 39655307 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01193675_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Traf3ip2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_134000.3 NP_598761.2 6 - 7 1752 140
GenBank mRNA AB238206.1 - 5 - 6 1331 140
AB238207.1 - 5 - 6 1331 140
AK031556.1 - 6 - 7 1477 140
AK037407.1 - 6 - 7 1559 140
AK052190.1 - 6 - 7 1752 140
AK098107.1 - 6 - 7 1474 140
BC008534.1 - 5 - 6 908 140
BC008535.1 - 5 - 6 908 140
BC022633.1 - 3 - 4 178 140
BC071207.1 - 3 - 4 246 140
BC096483.1 - 6 - 7 1433 5310588 140
BC138234.1 - 6 - 7 1522 8861253 140
BC138235.1 - 6 - 7 1522 8861254 140
JX869941.1 - 6 - 7 1759 140

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC022633.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Traf3ip2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

B cell apoptotic process
positive regulation of defense response to virus by host
humoral immune response
immunoglobulin secretion
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000002214.mm.1 NM_134000.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000002214.mm.1 NM_134000.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000002214.mm.1 TC1000000330.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0