This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192838_m1
See other SLC16A10 GE Assays ›
Gene Symbol:
Slc16a10
Gene Name:
solute carrier family 16 (monocarboxylic acid transporters), member 10
Gene Aliases:
2610103N14Rik, 9830169E08, Mct10, PRO0813, TAT1
Chromosome Location:
Chr.10: 40033532 - 40142264 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192838_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc16a10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114332.1 NP_001107804.1 4 - 5 1298 61
GenBank mRNA AK036743.1 - 4 - 5 1299 61
AK144135.1 - 4 - 5 1301 61
AK151290.1 - 4 - 5 1299 61
AK151636.1 - 4 - 5 1300 61
AK151704.1 - 4 - 5 1303 61
AK165240.1 - 4 - 5 1299 61
BC052877.1 - 4 - 5 779 30046388 61

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002036.mm.1

Set Membership:

Amplicon length less than or equal to 70 Transporter Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
ion transmembrane transport
transmembrane transport
transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000014462.mm.1 NM_001114332.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014462.mm.1 NM_001114332.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000014462.mm.1 TC1000002036.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0