This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01192039_m1
See other RIMKLA GE Assays ›
Gene Symbol:
Rimkla
Gene Name:
ribosomal modification protein rimK-like family member A
Gene Aliases:
B930030J24, NAAGS, NAAGS-II, Rimk
Chromosome Location:
Chr.4: 119465279 - 119493153 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01192039_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rimkla
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_177572.4 NP_808240.2 2 - 3 537 69
GenBank mRNA AK047164.1 - 2 - 3 532 69
BC057371.1 - 2 - 3 518 6835897 69
BU611721.1 - 2 - 3 567 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003426.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rimkla

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

cellular protein modification process
nucleotide binding
catalytic activity
ATP binding
ligase activity
metal ion binding
N-acetyl-L-aspartate-L-glutamate ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400027846.mm.1 NM_177572.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027846.mm.1 NM_177572.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400027846.mm.1 TC0400003426.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0