This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191924_g1
See other DDX56 GE Assays ›
Gene Symbol:
Ddx56
Gene Name:
DEAD (Asp-Glu-Ala-Asp) box polypeptide 56
Gene Aliases:
2600001H07Rik, D11Ertd619e, Noh61
Chromosome Location:
Chr.11: 6257545 - 6267729 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191924_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ddx56
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026538.3 NP_080814.1 7 - 8 1019 51
GenBank mRNA AK011136.1 - 7 - 8 1029 51
AK076047.1 - 7 - 8 1037 51
AK076975.1 - 7 - 8 1019 51
AK077514.1 - 7 - 8 1032 51
AK145628.1 - 7 - 8 1060 51
AK159793.1 - 7 - 8 1023 51
AK169654.1 - 7 - 8 1034 51
BC018291.1 - 7 - 8 1013 4190167 51

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002166.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ddx56

Gene Ontology Categories:

Function(s) Process(es)

rRNA processing
biological_process
RNA secondary structure unwinding
ribosome biogenesis
nucleotide binding
nucleic acid binding
RNA binding
ATP-dependent RNA helicase activity
helicase activity
ATP binding
hydrolase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100021103.mm.1 NM_026538.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021103.mm.1 NM_026538.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100021103.mm.1 TC1100002166.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1100021102.mm.1 NM_026538.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100021102.mm.1 NM_026538.3 Refseq
Affymetrix Mouse Clariom S Array PSR1100021102.mm.1 TC1100002166.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0