This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191876_m1
See other PPIL4 GE Assays ›
Gene Symbol:
Ppil4
Gene Name:
peptidylprolyl isomerase (cyclophilin)-like 4
Gene Aliases:
3732410E19Rik, 3830425H19Rik, AI788954, AW146233, PPIase
Chromosome Location:
Chr.10: 7792891 - 7822563 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191876_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ppil4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026141.3 NP_080417.2 9 - 10 922 125
GenBank mRNA AK011443.1 - 9 - 10 894 125
AK012371.1 - 9 - 10 899 125
AK014406.2 - 895 125
AK014453.1 - 8 - 9 1447 125
AK017370.1 - 9 - 10 922 125
AK167168.1 - 9 - 10 887 125
AK167258.1 - 9 - 10 894 125
BC079912.1 - 9 - 10 884 30661438 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000000053.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ppil4

Panther Classification:

Molecular Function -

chaperone

Gene Ontology Categories:

Function(s) Process(es)

protein peptidyl-prolyl isomerization
protein folding
biological_process
regulation of phosphorylation of RNA polymerase II C-terminal domain
nucleotide binding
nucleic acid binding
RNA binding
peptidyl-prolyl cis-trans isomerase activity
isomerase activity
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000000429.mm.1 NM_026141.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000000429.mm.1 NM_026141.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000000429.mm.1 TC1000000053.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0