This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191500_m1
See other MORC2A GE Assays ›
Gene Symbol:
Morc2a
Gene Name:
microrchidia 2A
Gene Aliases:
8430403M08Rik, Zcwcc1
Chromosome Location:
Chr.11: 3649494 - 3690477 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191500_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Morc2a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001159288.1 NP_001152760.1 7 - 8 1145 67
NM_198162.1 NP_937805.1 6 - 7 472 67
GenBank mRNA AK173042.1 - 7 - 8 1145 67
BC059243.1 - 6 - 7 472 6417551 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100000021.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Morc2a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
zinc ion binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1100000290.mm.1 NM_001159288.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000290.mm.1 NM_001159288.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000290.mm.1 NM_198162.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000290.mm.1 NM_198162.1 Refseq
Affymetrix Mouse Clariom S Array PSR1100000290.mm.1 TC1100000021.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0