This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01191053_m1
See other 4930467K11RIK GE Assays ›
Gene Symbol:
4930467K11Rik
Gene Name:
RIKEN cDNA 4930467K11 gene
Gene Aliases:
-
Chromosome Location:
Chr.10: 57478383 - 57486353 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01191053_m1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 4930467K11Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_045353.1 1 - 2 172 74
GenBank mRNA AK015522.1 - 1 - 2 172 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY715070.1 - 1 - 2 169 Chr. 10 - 57478381 - 57486342 [-] on Build GRCm38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream predicted gene,
36595
Gm36595 -1.0mb Anti-sense Chr. 10 - 56431504 - 56475419 on NCBI Build GRCm38
Downstream serine incorporator 1 Serinc1 1500011D18Rik,
AI315070,
AIGP-2,
TMS-2,
Tde1l,
Tde2,
Tms2,
mKIAA1253
29.4kb Sense Chr. 10 - 57515774 - 57532529 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002194.mm.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Agilent Affymetrix ABI

Array Name Probe IDs Targets Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 246647 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 AF101779.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 AK033024.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 BC053016.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 BC080287.1 genbank_mrna_mouse
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 NM_019743.3 refseq_mrna_mouse_NM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 779058 XM_893519.3 refseq_mrna_mouse_XM
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 414308 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 962421 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 67000 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179896 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179896 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137951 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179896 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27160532 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08841006 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27160537 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08841003 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27160541 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P27160539 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P08806370 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000015380.mm.1 NR_045353.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015380.mm.1 NR_045353.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015380.mm.1 TC1000002194.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000015382.mm.1 NR_045353.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015382.mm.1 NR_045353.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015382.mm.1 TC1000002194.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000015383.mm.1 NR_045353.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015383.mm.1 NR_045353.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015383.mm.1 TC1000002194.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000015378.mm.1 NR_045353.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015378.mm.1 NR_045353.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000015378.mm.1 TC1000002194.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 556944 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0