This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190971_m1
See other 9330102E08RIK GE Assays ›
Gene Symbol:
9330102E08Rik
Gene Name:
RIKEN cDNA 9330102E08 gene
Gene Aliases:
-
Chromosome Location:
Chr.6: 128169700 - 128183803 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190971_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 9330102E08Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_077223.1 1 - 2 1065 92
GenBank mRNA AK033854.1 - 1 - 2 1065 92

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream tetraspanin 9 Tspan9 6720474K14Rik,
9430079M16Rik,
AU018597
-38.3kb Sense Chr. 6 - 127961396 - 128143598 on NCBI Build GRCm38
Downstream TEA domain family member 4 Tead4 ETFR-2,
Etfr2,
FR-19,
Rtef1,
TEAD-4,
TEF-3,
Tcf13r1,
Tef3,
Tefr,
Tefr1,
Tefr1a
43.3kb Sense Chr. 6 - 128227143 - 128300813 on NCBI Build GRCm38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003192.mm.1

Set Membership:

Amplicon length between 86 and 100 Mus musculus Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600024869.mm.1 NR_077223.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024869.mm.1 TC0600003192.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024870.mm.1 NR_077223.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600024870.mm.1 TC0600003192.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0