This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01190323_m1
See other C87436 GE Assays ›
Gene Symbol:
C87436
Gene Name:
expressed sequence C87436
Gene Aliases:
-
Chromosome Location:
Chr.6: 86437328 - 86470388 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01190323_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: C87436
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001243742.1 NP_001230671.1 1 - 2 149 67
GenBank mRNA BC031913.1 - 1 - 2 103 5376246 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC031913.1, NM_001243742.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600007695.mm.1 NM_001243741.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007695.mm.1 NM_001243741.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007695.mm.1 NM_001243742.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007695.mm.1 NM_001243742.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007695.mm.1 TC0600000989.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600007691.mm.1 NM_001243741.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007691.mm.1 NM_001243741.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007691.mm.1 NM_001243742.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007691.mm.1 NM_001243742.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600007691.mm.1 TC0600000989.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0