Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400030788.mm.1 |
NM_001256112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030788.mm.1 |
NM_001256112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030788.mm.1 |
NM_029759.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030788.mm.1 |
NM_029759.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030788.mm.1 |
TC0400003740.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0400030787.mm.1 |
NM_001256112.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030787.mm.1 |
NM_001256112.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030787.mm.1 |
NM_029759.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030787.mm.1 |
NM_029759.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400030787.mm.1 |
TC0400003740.mm.1 |
MTA TC |