This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188445_m1
See other MTFR1L GE Assays ›
Gene Symbol:
Mtfr1l
Gene Name:
mitochondrial fission regulator 1-like
Gene Aliases:
2410166I05Rik, AI480750, Fam54b
Chromosome Location:
Chr.4: 134525555 - 134535387 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188445_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mtfr1l
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256112.1 NP_001243041.1 1 - 2 194 70
GenBank mRNA AK010825.1 - 1 - 2 186 70
AK089141.1 - 1 - 2 186 70
AK145362.1 - 1 - 2 186 70
AK171703.1 - 1 - 2 186 70
BY094859.1 - 1 - 2 194 70

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK010825.1, AK089141.1, AK145362.1, AK171703.1, NM_001256112.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Mus musculus 1700 Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

mitochondrial fission
biological_process
aerobic respiration
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400030793.mm.1 NM_001256112.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030793.mm.1 NM_001256112.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030793.mm.1 NM_029759.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030793.mm.1 NM_029759.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400030793.mm.1 TC0400003740.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400030798.mm.1 NM_001256112.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030798.mm.1 NM_001256112.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400030798.mm.1 TC0400003740.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 867881 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0