This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01188048_gH
See other DMAP1 GE Assays ›
Gene Symbol:
Dmap1
Gene Name:
DNA methyltransferase 1-associated protein 1
Gene Aliases:
1500016M21Rik, DNMAP1, DNMTAP1, Dmtap1, mKIAA1425
Chromosome Location:
Chr.4: 117674681 - 117682321 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01188048_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmap1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_023178.2 NP_075667.1 4 - 5 610 99
GenBank mRNA AB221661.1 - 4 - 5 553 99
AF265229.1 - 4 - 5 553 99
AF438610.1 - 4 - 5 610 99
AK005270.1 - 5 - 6 650 99
AK012055.1 - 5 - 6 631 99
AK136976.1 - 3 - 4 846 99
BC002321.1 - 4 - 5 503 99
BC023257.1 - 4 - 5 599 99
BC045160.1 - 4 - 5 590 6309705 99

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003373.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dmap1

Panther Classification:

Molecular Function -

chromatin/chromatin-binding, or -regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
DNA repair
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
chromatin modification
regulation of growth
positive regulation of transcription factor import into nucleus
histone H4 acetylation
histone H2A acetylation
negative regulation of transcription, DNA-templated
RNA polymerase II repressing transcription factor binding
chromatin binding
transcription corepressor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400027072.mm.1 NM_023178.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400027072.mm.1 NM_023178.2 Refseq
Affymetrix Mouse Clariom S Array PSR0400027072.mm.1 TC0400003373.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0