This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187963_g1
See other LCK GE Assays ›
Gene Symbol:
Lck
Gene Name:
lymphocyte protein tyrosine kinase
Gene Aliases:
Hck-3, Lsk, Lskt, p56<lck>, p56Lck
Chromosome Location:
Chr.4: 129548344 - 129573641 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187963_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lck
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162432.1 NP_001155904.1 5 - 6 463 82
NM_001162433.1 NP_001155905.1 5 - 6 505 82
NM_010693.3 NP_034823.1 4 - 5 560 82
GenBank mRNA AK088001.1 - 4 - 5 561 82
BC011474.1 - 5 - 6 470 4191610 82
BQ563042.1 - 5 - 6 454 82
M12056.1 - 4 - 5 536 82
X03533.1 - 4 - 5 566 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lck

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
activation of cysteine-type endopeptidase activity involved in apoptotic process
cell surface receptor signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
response to mechanical stimulus
response to metal ion
positive regulation of gene expression
phosphorylation
dephosphorylation
peptidyl-tyrosine phosphorylation
T cell differentiation
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
response to drug
positive regulation of tyrosine phosphorylation of Stat5 protein
response to hydrogen peroxide
regulation of apoptotic process
innate immune response
positive regulation of gamma-delta T cell differentiation
protein autophosphorylation
B cell receptor signaling pathway
regulation of T cell receptor signaling pathway
positive regulation of T cell activation
release of sequestered calcium ion into cytosol
positive regulation of uterine smooth muscle contraction
cellular response to peptide hormone stimulus
positive regulation of intrinsic apoptotic signaling pathway
nucleotide binding
glycoprotein binding
antigen binding
protein kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
protein serine/threonine phosphatase activity
receptor binding
protein binding
ATP binding
protein C-terminus binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein phosphatase binding
protein complex binding
SH2 domain binding
CD4 receptor binding
CD8 receptor binding
identical protein binding
phosphatidylinositol 3-kinase binding
ATPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_001162432.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_001162432.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_001162433.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_001162433.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_010693.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 NM_010693.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400029370.mm.1 TC0400003602.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-655dcf6cc8-78hlp:80/100.66.128.142:80.
git-commit: 0d764ad91c70359c551b4d51a80efc6847a0ee54
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0