This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187548_g1
See other ARHGAP4 GE Assays ›
Gene Symbol:
Arhgap4
Gene Name:
Rho GTPase activating protein 4
Gene Aliases:
A130009C12Rik, A530015A20Rik, Rgc1, c1, mKIAA0131
Chromosome Location:
Chr.X: 73894352 - 73911346 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187548_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Arhgap4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001162423.1 NP_001155895.1 6 - 7 908 72
NM_001162424.1 NP_001155896.1 5 - 6 845 72
NM_138630.2 NP_619536.2 6 - 7 908 72
GenBank mRNA AF443826.1 - 6 - 7 839 72
AK129060.1 - 1323 72
AK157294.1 - 6 - 7 907 72
AY183124.1 - 6 - 7 881 72
AY183125.1 - 5 - 6 818 72
AY183126.1 - 6 - 7 881 72
AY183127.1 - 6 - 7 881 72
AY183129.1 - 6 - 7 881 72
AY183133.1 - 5 - 6 818 72
BC109001.1 - 6 - 7 839 40048081 72
BC109002.1 - 5 - 6 776 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00003430.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Arhgap4

Panther Classification:

Molecular Function -

GTPase-activating protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of fibroblast migration
negative regulation of cell migration
negative regulation of axon extension
positive regulation of GTPase activity
GTPase activator activity
Rac GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_001162423.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_001162423.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_001162424.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_001162424.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_138630.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 NM_138630.2 Refseq
Affymetrix Mouse Clariom S Array PSR0X00015224.mm.1 TC0X00003430.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0