Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_001128606.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_001128606.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_001128607.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_001128607.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_183428.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
NM_183428.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0400029914.mm.1 |
TC0400003653.mm.1 |
MTA TC |