This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187457_m1
See other EPB4.1 GE Assays ›
Gene Symbol:
Epb4.1
Gene Name:
erythrocyte protein band 4.1
Gene Aliases:
4.1R, AI415518, D4Ertd442e, Elp-1, Elp1, Epb41, mKIAA4056
Chromosome Location:
Chr.4: 131923413 - 132075579 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187457_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Epb4.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001128606.1 NP_001122078.1 10 - 11 1675 85
NM_001128607.1 NP_001122079.1 10 - 11 1585 85
NM_183428.3 NP_906273.3 10 - 11 2937 85
GenBank mRNA AB370261.1 - 9 - 10 1460 85
AK147266.1 - 5 - 6 631 85
AK147644.1 - 5 - 6 631 85
AK220462.1 - 10 - 11 1582 85
BC068138.1 - 10 - 11 1545 85
BC079875.1 - 10 - 11 1504 6844082 85
CN526385.1 - 4 - 5 519 85
L00919.1 - 11 - 12 3012 85
LK021332.1 - 8 - 9 830 85
LK021333.1 - 8 - 9 830 85

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Epb4.1

Gene Ontology Categories:

Function(s) Process(es)

regulation of cell shape
actin cytoskeleton organization
cortical actin cytoskeleton organization
positive regulation of protein binding
actin binding
structural molecule activity
calmodulin binding
1-phosphatidylinositol binding
cytoskeletal protein binding
spectrin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0