Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0X00015095.mm.1 |
NM_008323.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015095.mm.1 |
NM_008323.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015095.mm.1 |
TC0X00002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015087.mm.1 |
NM_008323.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015087.mm.1 |
NM_008323.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015087.mm.1 |
TC0X00002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015086.mm.1 |
NM_008323.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015086.mm.1 |
NM_008323.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015086.mm.1 |
TC0X00002434.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0X00015091.mm.1 |
NM_008323.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015091.mm.1 |
NM_008323.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0X00015091.mm.1 |
TC0X00002434.mm.1 |
MTA TC |