This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187201_g1
See other PTCH2 GE Assays ›
Gene Symbol:
Ptch2
Gene Name:
patched homolog 2
Gene Aliases:
ptc2
Chromosome Location:
Chr.4: 117095785 - 117115383 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187201_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptch2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008958.2 NP_032984.1 20 - 21 3251 92
GenBank mRNA AB010833.1 - 20 - 21 3251 92
AK042653.1 - 725 92
BC058397.1 - 20 - 21 3506 92

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400001276.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptch2

Gene Ontology Categories:

Function(s) Process(es)

cell fate determination
epidermis development
epidermal cell fate specification
hair cycle
skin development
positive regulation of epidermal cell differentiation
smoothened binding
hedgehog receptor activity
hedgehog family protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400009606.mm.1 NM_008958.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400009606.mm.1 TC0400001276.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0