Assay Details
Gene Symbol: 1700003M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_040647.1 | 3 - 4 | 416 | 99 | ||
NR_040648.1 | 2 - 3 | 216 | 99 | |||
GenBank mRNA | AA198428.1 | - | 2 - 3 | 227 | 99 | |
AK005651.1 | - | 3 - 4 | 417 | 99 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | RIKEN cDNA E330017L17 gene | E330017L17Rik | -52.2kb | Sense | Chr. 4 - 129906226 - 129908996 on NCBI Build GRCm38 | |
Downstream | adhesion G protein-coupled receptor B2 | Adgrb2 | Bai2 | 21.3kb | Sense | Chr. 4 - 129984870 - 130022633 on NCBI Build GRCm38 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2039537 | AK005651.1 | mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 265291 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 190823 | - | - |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 267527 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2039537 | AK005651.1 | mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P101844 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299397 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299339 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299390 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299316 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03495375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03857733 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03841406 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299338 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03841410 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299340 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299399 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299309 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299400 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299334 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299357 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03852657 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299362 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299382 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03419162 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03555679 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299326 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299317 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04016701 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299319 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03841405 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299356 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299346 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03289343 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299395 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654877 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299292 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654862 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654878 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299372 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299305 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299368 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03246231 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654879 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299306 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03555680 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299353 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299394 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03758036 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03654875 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299376 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03758034 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04016697 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547297 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03852660 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299364 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03673325 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04016702 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03301171 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03814050 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299375 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547296 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547293 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03631286 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299370 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04016703 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03852661 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299393 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299327 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299351 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299344 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299371 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03814054 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P04016805 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03967532 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03857729 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03547303 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03631288 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03695295 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P03857735 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299318 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P23299407 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Clariom S Array | PSR0400011948.mm.1 | NR_040647.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011948.mm.1 | NR_040647.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011948.mm.1 | NR_040648.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011948.mm.1 | NR_040648.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011948.mm.1 | TC0400001537.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011946.mm.1 | NR_040647.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011946.mm.1 | NR_040647.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011946.mm.1 | NR_040648.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011946.mm.1 | NR_040648.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011946.mm.1 | TC0400001537.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0400011947.mm.1 | NR_040647.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011947.mm.1 | NR_040647.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011947.mm.1 | NR_040648.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011947.mm.1 | NR_040648.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0400011947.mm.1 | TC0400001537.mm.1 | MTA TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 419948 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
MouseWG-6 v2 Expression BeadChip | 105340193 | - | - |
