This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01187166_g1
See other 1700003M07RIK GE Assays ›
Gene Symbol:
1700003M07Rik
Gene Name:
RIKEN cDNA 1700003M07 gene
Gene Aliases:
-
Chromosome Location:
Chr.4: 129960374 - 129965138 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01187166_g1
Size
Availability Made To Order
Catalog # 4426961
Price 373.00
Your Price
Online offer:
373.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: 1700003M07Rik
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_040647.1 2 - 3 329 81
GenBank mRNA AK005651.1 - 2 - 3 330 81
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BY705842.1 - 2 - 3 327 Chr. 4 - 129960376 - 129963520 [+] on Build GRCm38 81

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream RIKEN cDNA E330017L17 gene E330017L17Rik -51.7kb Sense Chr. 4 - 129906226 - 129908996 on NCBI Build GRCm38
Downstream adhesion G protein-coupled receptor B2 Adgrb2 Bai2 23.6kb Sense Chr. 4 - 129984870 - 130022633 on NCBI Build GRCm38

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0