This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185476_m1
See other SMAP2 GE Assays ›
Gene Symbol:
Smap2
Gene Name:
small ArfGAP 2
Gene Aliases:
1810031K02Rik, AA408614, Smap1l
Chromosome Location:
Chr.4: 120968317 - 121017289 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185476_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smap2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_133716.3 NP_598477.2 6 - 7 963 70
GenBank mRNA AK152757.1 - 6 - 7 930 70
BC031581.1 - 6 - 7 952 70
BC032217.1 - 6 - 7 939 70
BC052413.1 - 6 - 7 963 5720177 70

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400003448.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smap2

Gene Ontology Categories:

Process(es)

GTPase activator activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400028010.mm.1 NM_133716.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028010.mm.1 NM_133716.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400028010.mm.1 TC0400003448.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400028009.mm.1 NM_133716.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400028009.mm.1 NM_133716.3 Refseq
Affymetrix Mouse Clariom S Array PSR0400028009.mm.1 TC0400003448.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0