This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01185366_m1
See other EYA4 GE Assays ›
Gene Symbol:
Eya4
Gene Name:
eyes absent 4 homolog (Drosophila)
Gene Aliases:
B130023L16Rik
Chromosome Location:
Chr.10: 23102962 - 23350856 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01185366_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eya4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010167.4 NP_034297.2 5 - 6 507 116
GenBank mRNA AK041625.1 - 5 - 6 508 116
BC120898.1 - 5 - 6 491 40061597 116
BC120899.1 - 5 - 6 491 40061599 116
Y17115.1 - 6 - 7 618 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000001868.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eya4

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
transcription, DNA-templated
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
multicellular organism development
sensory perception of sound
chromatin modification
middle ear morphogenesis
molecular_function
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
hydrolase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000013491.mm.1 NM_010167.5 Refseq
Affymetrix Mouse Clariom S Array PSR1000013491.mm.1 TC1000001868.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000013492.mm.1 NM_010167.5 Refseq
Affymetrix Mouse Clariom S Array PSR1000013492.mm.1 TC1000001868.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0