This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184862_g1
See other LOXL3 GE Assays ›
Gene Symbol:
Loxl3
Gene Name:
lysyl oxidase-like 3
Gene Aliases:
Lor2, Loxl2
Chromosome Location:
Chr.6: 83034224 - 83052564 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184862_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Loxl3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_013586.4 NP_038614.2 12 - 13 2178 77
GenBank mRNA AF053368.1 - 12 - 13 2209 77
AK030548.1 - 12 - 13 2179 77
AK145189.1 - 7 - 8 1352 77
BC011298.1 - 12 - 13 2172 3488978 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000914.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Loxl3

Panther Classification:

Molecular Function -

oxidase

Gene Ontology Categories:

Function(s) Process(es)

epithelial to mesenchymal transition
negative regulation of transcription, DNA-templated
oxidation-reduction process
scavenger receptor activity
copper ion binding
aspartate oxidase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-NH2 group of donors, oxygen as acceptor
D-lysine oxidase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header