This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184720_m1
See other EMC3 GE Assays ›
Gene Symbol:
Emc3 Cyfip1
Gene Name:
ER membrane protein complex subunit 3
cytoplasmic FMR1 interacting protein 1
Gene Aliases:
0610039A15Rik, AI225901, AW260416, Pob, Tmem111
E030028J09Rik, P140SRA-1, P140sra1, Shyc, Sra-1, Sra1, l(7)1Rl, l71Rl, l7Rl1, mKIAA0068, pl-1
Chromosome Location:
Chr.6: 113514887 - 113531638 on Build GRCm38
Chromosome Location:
Chr.7: 55842071 - 55932633 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
-
Assay ID Mm01184720_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Emc3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_175101.3 NP_780310.1 5 - 6 651 64
GenBank mRNA AK075595.1 - 5 - 6 652 64
AK167819.1 - 5 - 6 500 64
AK200024.1 - 2 - 3 104 64
AK210449.1 - 5 - 6 361 64
AK218757.1 - 2 - 3 104 64
BC004641.1 - 5 - 6 647 3157147 64
Gene Symbol: Cyfip1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK205520.1 - 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002924.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Emc3
Click here to find siRNAs for Gene Cyfip1

Gene Ontology Categories:

Function(s) Process(es)

biological_process
protein folding in endoplasmic reticulum
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600022472.mm.1 NM_175101.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600022472.mm.1 NM_175101.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600022472.mm.1 TC0600002924.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0