This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184572_g1
See other OGG1 GE Assays ›
Gene Symbol:
Ogg1
Gene Name:
8-oxoguanine DNA-glycosylase 1
Gene Aliases:
Mmh
Chromosome Location:
Chr.6: 113326976 - 113334188 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184572_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ogg1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010957.4 NP_035087.3 6 - 7 1197 109
GenBank mRNA AF000669.1 - 6 - 7 1075 109
AF003596.1 - 6 - 7 1162 109
AK008192.1 - 225 109
AK146870.1 - 6 - 7 1197 109
AK165875.1 - 6 - 7 1150 109
AK167456.1 - 6 - 7 1257 109
BC138733.1 - 6 - 7 1153 8861755 109
BC138736.1 - 6 - 7 1153 8861758 109
U88621.1 - 6 - 7 1165 109
Y11247.1 - 6 - 7 1128 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001246.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ogg1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA repair
base-excision repair
nucleotide-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
metabolic process
response to radiation
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
negative regulation of apoptotic process
nucleic acid phosphodiester bond hydrolysis
negative regulation of double-strand break repair via single-strand annealing
purine-specific mismatch base pair DNA N-glycosylase activity
oxidized base lesion DNA N-glycosylase activity
DNA binding
damaged DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Agilent Affymetrix Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758829 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758831 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758832 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758833 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489903 U96711.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P489903 U96711.1 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 857816 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 397374 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767851 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 302727 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 969859 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 29699 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133178 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489903 U96711.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P489903 U96711.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489903 U96711.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P489903 U96711.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P567905 U96711.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758836 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758834 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758819 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758837 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758827 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758830 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758849 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758841 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758833 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758847 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758823 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758821 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758844 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758822 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758829 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758824 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758848 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24758851 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133178 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06030660 - -
Whole Mouse Genome G4122A A_51_P489903 U96711.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P489903 U96711.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P567905 U96711.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P103503 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133178 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05606174 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05060534 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05060535 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05606175 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P05606183 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758833 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758832 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758829 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24758831 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758911 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758824 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758934 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758910 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758922 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758942 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758920 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758872 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758848 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758823 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758833 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758874 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758849 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758914 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758857 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758932 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758904 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758936 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758940 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758822 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758915 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758901 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758852 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758931 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758847 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758917 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758888 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758859 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758897 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758819 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758899 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758864 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758889 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758851 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758918 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758925 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758825 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758926 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758867 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758924 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758827 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758844 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758930 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758908 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758870 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758830 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758938 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758829 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758865 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758891 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758916 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758893 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758863 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758941 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758860 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758853 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758862 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758944 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758836 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758906 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758839 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758841 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758913 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758927 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24758855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758830 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05433975 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05606180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758874 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758867 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758852 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758860 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758827 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05449530 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05267433 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05019232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05433979 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758825 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758823 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758821 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05331180 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758873 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758859 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758864 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758853 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758865 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758870 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758844 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758863 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05606175 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758849 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758847 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758857 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758866 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758819 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758855 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758833 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758839 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P24758841 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10540644 - -
Affymetrix Mouse Gene 1.0 ST Array 10540647 - -
Affymetrix Mouse Gene 1.0 ST Array 10540648 - -
Affymetrix Mouse Gene 1.0 ST Array 10540642 - -
Affymetrix Mouse Gene 1.0 ST Array 10540643 - -
Affymetrix Mouse Gene 1.0 ST Array 10540641 - -
Affymetrix Mouse Gene 1.0 ST Array 10540640 - -
Affymetrix Mouse Gene 1.0 ST Array 10540646 - -
Affymetrix Mouse Gene 1.0 ST Array 10540645 - -
Affymetrix HT MG-430 PM Array Plate 1448815_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1430078_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10540639 - -
Mouse Genome 430 2.0 Array 1430078_a_at - -
Mouse Genome 430 2.0 Array 1448815_PM_at - -
Mouse Genome 430 2.0 Array 1448815_at - -
Mouse Genome 430 2.0 Array 1430078_PM_a_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18294_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 18294_at - -
Affymetrix Mouse Clariom S Array PSR0600009652.mm.1 NM_010957.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009652.mm.1 NM_010957.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600009652.mm.1 TC0600001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009653.mm.1 NM_010957.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009653.mm.1 NM_010957.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600009653.mm.1 TC0600001246.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600009654.mm.1 NM_010957.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600009654.mm.1 NM_010957.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600009654.mm.1 TC0600001246.mm.1 MTA TC
Mouse Expression Set 430 1448815_at - -
Mouse Expression Set 430 1430078_a_at - -
Mouse Expression Set 430 1448815_PM_at - -
Mouse Expression Set 430 1430078_PM_a_at - -
Murine Genome U74v2 Set 160882_at - -
Murine Genome U74v2 Set 160882_PM_at - -
Murine Genome U74v2 Set 165929_PM_at - -
Murine Genome U74v2 Set 165929_at - -
Mouse Genome 430A 2.0 Array 1430078_PM_a_at - -
Mouse Genome 430A 2.0 Array 1448815_PM_at - -
Mouse Genome 430A 2.0 Array 1430078_a_at - -
Mouse Genome 430A 2.0 Array 1448815_at - -
Affymetrix HT Mouse Genome 430A Array 1448815_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1430078_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1430078_a_at - -
Affymetrix HT Mouse Genome 430A Array 1448815_at - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 106370142 - -
Illumina mouse-6 v1.0 expression beadchip 1940279 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001940279 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2550230 - -
mouse-6 v1.1 (Illumina) scl29634.7.1_101-S - -
Illumina Sentrix mouse-6.v1 bead array 106370142 - -
Illumina Sentrix mouse-6.v1 bead array 1940279 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl29634.7.1_101-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 106370142 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1940279 - -
MouseRef-8 v2 Expression BeadChip ILMN_2991864 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2550230 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2991864 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241289 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0