This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01184277_g1
See other IQSEC3 GE Assays ›
Gene Symbol:
Iqsec3
Gene Name:
IQ motif and Sec7 domain 3
Gene Aliases:
AK122446, BRAG3, mKIAA1110, synarfGEF
Chromosome Location:
Chr.6: 121372927 - 121473703 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01184277_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Iqsec3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033354.3 NP_001028526.1 6 - 7 2405 59
NM_001289685.1 NP_001276614.1 5 - 6 2134 59
NR_110358.1 5 - 6 2134 59
GenBank mRNA AK122446.1 - 5 - 6 2121 59
AK147351.1 - 5 - 6 2080 59
AK169437.1 - 6 - 7 2405 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003060.mm.1

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

actin cytoskeleton organization
regulation of ARF protein signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
activation of GTPase activity
ARF guanyl-nucleotide exchange factor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600023661.mm.1 NM_001033354.3 Refseq
Affymetrix Mouse Clariom S Array PSR0600023661.mm.1 NM_001289685.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023661.mm.1 NR_110358.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600023661.mm.1 TC0600003060.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0