This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183641_g1
See other FAM136A GE Assays ›
Gene Symbol:
Fam136a
Gene Name:
family with sequence similarity 136, member A
Gene Aliases:
2010309E21Rik, AW124904, Hyccin
Chromosome Location:
Chr.6: 86365453 - 86370063 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183641_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Fam136a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025591.2 NP_079867.1 1 - 2 136 83
GenBank mRNA AK005280.1 - 1 - 2 96 83
AK008550.1 - 1 - 2 102 83
AK141279.1 - 1 - 2 96 83
BC013621.1 - 1 - 2 84 83
BC024049.1 - 1 - 2 136 5100000 83
BC103775.1 - 1 - 2 137 6485116 83
CT010180.1 - 1 - 2 86 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Fam136a

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0