This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183596_m1
See other PCYOX1 GE Assays ›
Gene Symbol:
Pcyox1
Gene Name:
prenylcysteine oxidase 1
Gene Aliases:
1200015P13Rik, AI115532, AI448340, PCL1, Pcly
Chromosome Location:
Chr.6: 86386006 - 86397158 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183596_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pcyox1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_025823.4 NP_080099.1 1 - 2 176 126
GenBank mRNA AK004799.2 - 1 - 2 131 126
AK004840.1 - 1 - 2 131 126
AK031585.1 - 1 - 2 132 126
AK033373.1 - 1 - 2 132 126
AK049412.1 - 1 - 2 129 126
AK134528.1 - 1 - 2 177 126
AK145366.1 - 1 - 2 136 126
AK147190.1 - 1 - 2 144 126
AK159981.1 - 1 - 2 132 126
AK161616.1 - 1 - 2 132 126
AK169209.1 - 1 - 2 132 126
AK173058.1 - 1 - 2 98 126
BC028308.1 - 1 - 2 146 3486530 126

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600002635.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pcyox1

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

chloride transport
prenylated protein catabolic process
prenylcysteine catabolic process
oxidation-reduction process
chloride transmembrane transport
prenylcysteine oxidase activity
chloride-transporting ATPase activity
oxidoreductase activity
oxidoreductase activity, acting on a sulfur group of donors, oxygen as acceptor

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600020459.mm.1 NM_025823.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020459.mm.1 NM_025823.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600020459.mm.1 TC0600002635.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600020457.mm.1 NM_025823.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600020457.mm.1 NM_025823.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600020457.mm.1 TC0600002635.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 632833 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header