This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01183194_g1
See other SLC25A18 GE Assays ›
Gene Symbol:
Slc25a18
Gene Name:
solute carrier family 25 (mitochondrial carrier), member 18
Gene Aliases:
1500015I14Rik, AW125787
Chromosome Location:
Chr.6: 120773510 - 120794169 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01183194_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Slc25a18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001081048.2 NP_001074517.1 7 - 8 661 100
GenBank mRNA AK005250.1 - 5 - 6 573 100
BC116828.1 - 7 - 8 661 40126147 100
BC137603.1 - 7 - 8 661 8860622 100

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

transporter secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
biological_process
transmembrane transport
molecular_function
symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600010806.mm.1 NM_001081048.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600010806.mm.1 NM_001081048.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600010806.mm.1 TC0600001354.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0