Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600023538.mm.1 |
NM_007510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023538.mm.1 |
NM_007510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023538.mm.1 |
TC0600003045.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023535.mm.1 |
NM_007510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023535.mm.1 |
NM_007510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023535.mm.1 |
TC0600003045.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023536.mm.1 |
NM_007510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023536.mm.1 |
NM_007510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023536.mm.1 |
TC0600003045.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600023537.mm.1 |
NM_007510.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023537.mm.1 |
NM_007510.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600023537.mm.1 |
TC0600003045.mm.1 |
MTA TC |