This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01182929_m1
See other TNFRSF1A GE Assays ›
Gene Symbol:
Tnfrsf1a
Gene Name:
tumor necrosis factor receptor superfamily, member 1a
Gene Aliases:
CD120a, FPF, TNF-R, TNF-R-I, TNF-R1, TNF-R55, TNF-alphaR1, TNFAR, TNFR60, TNFRI, TNFRp55, TNFalpha-R1, Tnfr-2, Tnfr1, p55, p55-R
Chromosome Location:
Chr.6: 125349723 - 125362483 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01182929_m1
Size
Availability Inventoried
Catalog # 4331182
Price 244.00
Your Price
Online offer: 200.00
200.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf1a
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011609.4 NP_035739.2 5 - 6 855 82
GenBank mRNA AK154388.1 - 5 - 6 825 82
AK159989.1 - 5 - 6 82
AK178992.1 - 3 - 4 326 82
AK206708.1 - 3 - 4 348 82
AY541589.1 - 5 - 6 536 82
AY541590.1 - 5 - 6 536 82
BC004599.1 - 5 - 6 771 3585060 82
BC052675.1 - 5 - 6 790 30061894 82
L26349.1 - 5 - 6 823 82
M59377.1 - 5 - 6 649 82
M60468.1 - 5 - 6 748 82
X57796.1 - 5 - 6 826 82
X59238.1 - 5 - 6 748 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600001459.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf1a

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin metabolic process
apoptotic process
defense response
inflammatory response
signal transduction
cell surface receptor signaling pathway
intrinsic apoptotic signaling pathway in response to DNA damage
positive regulation of gene expression
negative regulation of gene expression
programmed cell death
cytokine-mediated signaling pathway
negative regulation of interleukin-6 production
positive regulation of tumor necrosis factor production
positive regulation of protein import into nucleus, translocation
tumor necrosis factor-mediated signaling pathway
positive regulation of tyrosine phosphorylation of Stat1 protein
defense response to bacterium
regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of neuron apoptotic process
positive regulation of angiogenesis
positive regulation of transcription from RNA polymerase II promoter
negative regulation of inflammatory response
positive regulation of inflammatory response
protein heterooligomerization
protease binding
tumor necrosis factor-activated receptor activity
protein binding
protein complex binding
tumor necrosis factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0