This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181889_g1
See other SEMA4F GE Assays ›
Gene Symbol:
Sema4f
Gene Name:
sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain
Gene Aliases:
Sema W
Chromosome Location:
Chr.6: 82911884 - 82939761 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181889_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB021291.1 - 8 - 9 1101 71
AF038652.1 - 7 - 8 860 71
AK035993.1 - 8 - 9 1133 71
AK047067.1 - 8 - 9 1117 71
AK143196.1 - 8 - 9 1145 71
AK158292.1 - 8 - 9 71
BC057048.1 - 7 - 8 1009 71
BC090625.1 - 8 - 9 1103 71
BC094567.1 - 8 - 9 1128 6834138 71
BC139232.1 - 8 - 9 8862254 71

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7db4fb6694-gx648:80/100.66.128.142:80.
git-commit: 672a5a8c50d6b616e949be631a941c86748ade1b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header