This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181884_g1
See other SEMA4F GE Assays ›
Gene Symbol:
Sema4f
Gene Name:
sema domain, immunoglobulin domain (Ig), TM domain, and short cytoplasmic domain
Gene Aliases:
Sema W
Chromosome Location:
Chr.6: 82911884 - 82939761 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181884_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Sema4f
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AB021291.1 - 3 - 4 454 82
AF038652.1 - 2 - 3 213 82
AK035993.1 - 3 - 4 486 82
AK047067.1 - 3 - 4 470 82
AK143196.1 - 3 - 4 497 82
AK158292.1 - 3 - 4 420 82
BC057048.1 - 3 - 4 456 82
BC090625.1 - 3 - 4 456 82
BC094567.1 - 3 - 4 481 6834138 82
BC139232.1 - 3 - 4 428 8862254 82

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

membrane-bound signaling molecule intercellular signal molecule

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600019759.mm.1 NM_011350.4 Refseq
Affymetrix Mouse Clariom S Array PSR0600019759.mm.1 TC0600002553.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0