This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181435_m1
See other MMRN1 GE Assays ›
Gene Symbol:
Mmrn1
Gene Name:
multimerin 1
Gene Aliases:
4921530G03Rik, Emilin4, Mmrn
Chromosome Location:
Chr.6: 60944317 - 60989378 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181435_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmrn1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163507.1 NP_001156979.1 3 - 4 1051 88
NM_027613.1 NP_081889.1 3 - 4 1051 88
GenBank mRNA AK036724.1 - 3 - 4 889 88
AK137556.1 - 3 - 4 890 88
BC137623.1 - 3 - 4 851 8860642 88
BC144789.1 - 3 - 4 851 9053337 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600000724.mm.1

Set Membership:

Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600005558.mm.1 NM_001163507.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005558.mm.1 NM_001163507.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005558.mm.1 NM_027613.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005558.mm.1 NM_027613.1 Refseq
Affymetrix Mouse Clariom S Array PSR0600005558.mm.1 TC0600000724.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0