This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181365_m1
See other CAMK2B GE Assays ›
Gene Symbol:
Camk2b
Gene Name:
calcium/calmodulin-dependent protein kinase II, beta
Gene Aliases:
-
Chromosome Location:
Chr.11: 5969642 - 6065748 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181365_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Camk2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001174053.1 NP_001167524.1 3 - 4 429 80
NM_001174054.1 NP_001167525.1 3 - 4 429 80
NM_007595.5 NP_031621.3 3 - 4 429 80
GenBank mRNA AK046454.1 - 3 - 4 430 80
AK163698.1 - 3 - 4 246 80
BC024803.1 - 3 - 4 245 80
BC080273.1 - 3 - 4 231 6851995 80

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1100002158.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Protein Phosphatase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Camk2b

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
inhibitory G-protein coupled receptor phosphorylation
protein phosphorylation
calcium ion transport
nervous system development
positive regulation of neuron projection development
phosphorylation
peptidyl-serine phosphorylation
cell differentiation
regulation of synaptic transmission, cholinergic
positive regulation of phospholipase A2 activity
response to cadmium ion
protein autophosphorylation
regulation of long-term neuronal synaptic plasticity
cell projection morphogenesis
neuromuscular process controlling balance
long-term synaptic potentiation
activation of meiosis involved in egg activation
positive regulation of dendritic spine morphogenesis
positive regulation of synapse maturation
positive regulation of apoptotic signaling pathway
nucleotide binding
protein kinase activity
protein serine/threonine kinase activity
calmodulin-dependent protein kinase activity
protein binding
calmodulin binding
ATP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein homodimerization activity
phospholipase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0