Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1100001007.mm.1 |
NM_001005867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001007.mm.1 |
NM_001005867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001007.mm.1 |
NM_028601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001007.mm.1 |
TC1100000094.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1100001006.mm.1 |
NM_001005867.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001006.mm.1 |
NM_001005867.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001006.mm.1 |
NM_028601.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1100001006.mm.1 |
TC1100000094.mm.1 |
MTA TC |