This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01181016_m1
See other EPHB2 GE Assays ›
Gene Symbol:
Ephb2
Gene Name:
Eph receptor B2
Gene Aliases:
Cek5, Drt, ETECK, Erk, Hek5, Nuk, Prkm5, Qek5, Sek3, Tyro5
Chromosome Location:
Chr.4: 136647536 - 136836295 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01181016_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ephb2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290753.1 NP_001277682.1 2 - 3 309 74
NM_010142.3 NP_034272.1 2 - 3 309 74
GenBank mRNA BC043088.1 - 2 - 3 233 6418088 74
BC062924.1 - 2 - 3 254 74
CJ067955.1 - 2 - 3 309 74
L25890.1 - 3 - 4 151 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ephb2

Panther Classification:

Molecular Function -

transmembrane signal receptor

Gene Ontology Categories:

Function(s) Process(es)

cell morphogenesis
angiogenesis
urogenital system development
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
multicellular organism development
nervous system development
axon guidance
axonal fasciculation
learning
organ morphogenesis
phosphorylation
peptidyl-tyrosine phosphorylation
optic nerve morphogenesis
central nervous system projection neuron axonogenesis
corpus callosum development
retinal ganglion cell axon guidance
inner ear morphogenesis
ephrin receptor signaling pathway
regulation of neuronal synaptic plasticity
positive regulation of long-term neuronal synaptic plasticity
camera-type eye morphogenesis
regulation of axonogenesis
negative regulation of axonogenesis
regulation of body fluid levels
positive regulation of synapse assembly
palate development
dendritic spine development
dendritic spine morphogenesis
commissural neuron axon guidance
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor activity
ephrin receptor activity
transmembrane-ephrin receptor activity
receptor binding
protein binding
ATP binding
axon guidance receptor activity
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0