Assay Details
Gene Symbol: Ccdc33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029212.3 | NP_083488.1 | 1 - 2 | 304 | 82 | |
GenBank mRNA | AB048860.1 | - | 1 - 2 | 378 | 82 | |
AK015980.1 | - | 1 - 2 | 378 | 82 | ||
BC116291.1 | - | 1 - 2 | 304 | 40094912 | 82 | |
BC116292.1 | - | 1 - 2 | 304 | 40094918 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AB048860.1, AK015980.1, BC116291.1, BC116292.1, NM_029212.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
