This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01178676_m1
See other CDON GE Assays ›
Gene Symbol:
Cdon
Gene Name:
cell adhesion molecule-related/down-regulated by oncogenes
Gene Aliases:
CDO, Cd0, Orcam
Chromosome Location:
Chr.9: 35421129 - 35507652 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01178676_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cdon
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_021339.2 NP_067314.2 10 - 11 2171 77
GenBank mRNA AF090866.1 - 10 - 11 2171 77
BC109176.2 - 10 - 11 2170 40048829 77

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900000336.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cdon

Gene Ontology Categories:

Function(s) Process(es)

cell fate specification
positive regulation of protein phosphorylation
lens development in camera-type eye
cell adhesion
smoothened signaling pathway
myoblast fusion
anterior/posterior pattern specification
embryonic body morphogenesis
skeletal muscle satellite cell differentiation
regulation of striated muscle tissue development
single organismal cell-cell adhesion
cerebral cortex development
positive regulation of MAPK cascade
regulation of protein heterodimerization activity
positive regulation of myoblast differentiation
regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic morphogenesis
positive regulation of skeletal muscle tissue development
positive regulation of small GTPase mediated signal transduction
striated muscle cell differentiation
embryonic retina morphogenesis in camera-type eye
positive regulation of neural precursor cell proliferation
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900002504.mm.1 NM_021339.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900002504.mm.1 NM_021339.2 Refseq
Affymetrix Mouse Clariom S Array PSR0900002504.mm.1 TC0900000336.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0