This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm01178269_m1
See other DPY19L2 GE Assays ›
Gene Symbol:
Dpy19l2
Gene Name:
dpy-19-like 2 (C. elegans)
Gene Aliases:
4932443J21Rik, Gm18376
Chromosome Location:
Chr.9: 24557047 - 24696941 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm01178269_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dpy19l2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001166207.1 NP_001159679.1 3 - 4 621 75
GenBank mRNA AK030128.1 - 3 - 4 611 75

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0900001923.mm.1

Set Membership:

Amplicon length between 71 and 85 Mus musculus Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

multicellular organism development
spermatogenesis
spermatid development
protein C-linked glycosylation via 2'-alpha-mannosyl-L-tryptophan
cell differentiation
alpha-1,6-mannosyltransferase activity
alpha-1,2-mannosyltransferase activity
mannosyltransferase activity
alpha-1,3-mannosyltransferase activity
alpha-1,3-galactosyltransferase activity
molecular_function
UDP-glucose:glycoprotein glucosyltransferase activity
glycolipid mannosyltransferase activity
oligosaccharyl transferase activity
dolichyl-phosphate-glucose-glycolipid alpha-glucosyltransferase activity
acetylglucosaminyltransferase activity
acetylgalactosaminyltransferase activity
galactosyltransferase activity
fucosyltransferase activity
O antigen polymerase activity
lipopolysaccharide-1,6-galactosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
cellulose synthase activity
9-phenanthrol UDP-glucuronosyltransferase activity
1-phenanthrol glycosyltransferase activity
9-phenanthrol glycosyltransferase activity
1,2-dihydroxy-phenanthrene glycosyltransferase activity
phenanthrol glycosyltransferase activity
beta-1,4-mannosyltransferase activity
alpha-1,2-galactosyltransferase activity
dolichyl pyrophosphate Man7GlcNAc2 alpha-1,3-glucosyltransferase activity
lipopolysaccharide-1,5-galactosyltransferase activity
dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase activity
inositol phosphoceramide synthase activity
glucosyltransferase activity
alpha-(1->3)-fucosyltransferase activity
alpha-(1->6)-fucosyltransferase activity
indole-3-butyrate beta-glucosyltransferase activity
salicylic acid glucosyltransferase (ester-forming) activity
salicylic acid glucosyltransferase (glucoside-forming) activity
benzoic acid glucosyltransferase activity
chondroitin hydrolase activity
dolichyl-pyrophosphate Man7GlcNAc2 alpha-1,6-mannosyltransferase activity
cytokinin 9-beta-glucosyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0900016179.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016179.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016179.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016174.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016174.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016174.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016176.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016176.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016176.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016177.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016177.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016177.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016173.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016173.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016173.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016172.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016172.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016172.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016175.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016175.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016175.mm.1 TC0900001923.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0900016178.mm.1 NM_001166207.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016178.mm.1 NM_001166207.1 Refseq
Affymetrix Mouse Clariom S Array PSR0900016178.mm.1 TC0900001923.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0